Quick Connect™ Swivels, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
QCS-D
89134-252EA 81.45 USD
89134-252
Quick Connect™ Swivels, SAI Infusion Technologies
Animal Supplies Animal Infusion Supplies
The Quick Connect™ Swivel provides Luer-Lock fittings for trouble-free connection to extension lines and tethers, making connecting simple, fast, and easy.
  • Fits all standard luer fittings
  • Double Luer and Single Luer models available
  • EtO exposed
Add injection caps, valves, or stopcocks to simplify dosing or sampling procedures. Part of SAI's Quick Connect™ System, which includes the Quick Connect™ Tether and Quick Connect™ Harness for tethered small animal infusion studies.
Packaging: Packaged individually and sterile.