QuickConnect™ Dual Luer Spring Tether, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
QCT-12
89134-250EA 19.44 USD
89134-250
QuickConnect™ Dual Luer Spring Tether, SAI Infusion Technologies
Animal Supplies Animal Infusion Supplies
The Dual Luer Spring Tether is a standard, stainless steel tether with a male luered polyurethane extension line built inside. The male luered fittings connect to the QuickConnect™ Harness and QuickConnect™ Swivel to complete the system.
  • Connections at the catheter and swivel ends are fast and easy
  • Compatible with any female Luer
  • Lined with polyurethane tubing
  • Length is customizable
This flexible stainless steel spring tether will protect the infusion line from damage due to biting during both short-term and long-term infusion studies. Easily connect to a QuickConnect™ swivel for freedom of movement without any tether twisting.
Packaging: Individually packaged and EtO exposed. Supplied sterile.