Quick Connect™ Jacket Adaptors, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
QCJA-23
89134-212EA 70.54 USD
89134-212
Quick Connect™ Jacket Adaptors, SAI Infusion Technologies
Animal Supplies Animal Restraints
The Quick Connect™ Jacket Adaptor is designed for researchers who prefer a woven rodent jacket over an elastomeric harness for continuous or intermittent infusion. The Jacket Adaptor utilizes the same luer design as the Quick Connect™ Harness, but connects securely to a cloth jacket with Velcro™, and can be adjusted as the animal grows.
  • Attached injection cap can be removed, or pierced with a Huber needle
  • Choose from two catheter connector pin sizes to fit 21G and 23G catheters
  • To be used with an SAI/Lomir Jacket
  • Fit can be adjusted as the animal grows
  • Individually packaged
  • EtO exposed
These jackets are part of SAI's Quick Connect™ System, which includes the Quick Connect™ Tether, Harness, and Swivel for tethered small animal infusion studies.
Packaging: Packaged individually and sterile.