Puritan® Spatula Shaped Knitted Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
3676
89133-806CS 380.52 USD
89133-806
Puritan® Spatula Shaped Knitted Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
Cleaning swab for use in critical environments
Spatula-shaped knitted polyester tip. Polypropylene handle. Low NVR extraction and particle generation. Nonsterile. Made in the USA.
Certifications: CE marked.
Ordering information: Packaged 100 per pack, 10 packs per case.