Puritan® Knitted Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
3135
89133-794CS 491.14 USD
89133-794 89133-804
Puritan® Knitted Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Clinical Use
These knitted polyester swabs are non-abrasive and lint-free
Ideal for instrumentation and medical device manufacturing. Made in the USA.
In addition, the polyester can easily stand up to IPA, acetone, and other solvents commonly used in clean rooms and other critical environments.
Certifications: CE marked.
Ordering information: Swabs are packaged 100 per pack, 10 packs per case.