PurSwab® Pointed Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
1130
89133-790CS 244.36 USD
89133-790
PurSwab® Pointed Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Clinical Use
Pointed foam tip is heat staked to a green polypropylene handle and fits in small tight areas
Made in the USA.
Used for precision cleaning. Can be used with IPA and DI water. Lint- and residue-free.
Certifications: CE marked.
Ordering information: Packaged as 10 bags if 100 swabs.