PurSwab® Small Paddle Shaped Foam Tipped Applicator, Black Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
1273BK
89133-786CS 201.33 USD
89133-786
PurSwab® Small Paddle Shaped Foam Tipped Applicator, Black Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Clinical Use
These swabs have thermal edge seals and thermal bonding to the handle help ensure the sponge-like foam will clean up unwanted particles without releasing any into your critical environment.
Small flat paddle that is lint and residue free.
Made in the USA.
Certifications: CE marked.
Ordering information: Packaged as 2 bags of 500 swabs per bag.