Corning® LSE™ Vortex Mixer, Corning
Supplier: Corning
6775
89133-512CS 994.14 USD
89133-512
Corning® LSE™ Vortex Mixer, Corning
Shakers and Mixers Vortex Mixers
The Corning variable speed LSE Vortex Mixer provides fast, efficient mixing with minimal vibration
The unit features a powerful, counter-balanced motor drive, continuous or "touch" mix modes. A range of accessories are available for all common sample tubes and containers.
Certifications: ETL Certified and CSA Compliant.