Corning® LSE™ High Speed Microcentrifuge, Corning
Supplier: Corning
6765-HS
89133-510CS 3394.88 USD
89133-510
Corning® LSE™ High Speed Microcentrifuge, Corning
Centrifuges Microcentrifuges
The Corning® LSE High Speed Microcentrifuge features an easy-to-use, digital control interface and high-speed performance for quick nucleic acid and protein separations
  • Fast acceleration to 13,300 rpm/16,300 x g
  • Quiet and cool running even at maxium speed
Two control knobs are used to precisely set rotor speed and run time, and parameters are shown on the easy-to-read LED displays. Press the "Speed/G-Force" knob to toggle the display between rpm and calculated g-force.
The LSE High Speed Microcentrifuge incorporates a brushless drive and unique air cooling system to both reduce noise and minimize sample heating even during prolonged runs at maximum speed.
Supplied with a 24 x 1.5/2.0 mL rotor for micro samples. Adapters are available for tubes smaller than 1.5 mL.