Gram color staining kit unstabilised
Supplier: VWR International
89133-285
89133-285KT 113.99 USD
89133-285
Gram color staining kit unstabilised
Gram color staining kit
Traditional Gram Stain Kit (unstabilized Iodine) for differentiating bacteria in cultures and clinical samples.
  • Set consists of "pre-mixed" iodine
  • For in vitro diagnostic use
  • Specifically formulatess for differential staining of bacteria from cultures or patient specimens
Kit includes one 8oz. bottle each crystal violet, decolorizer, safranin, and 4oz. unstabilized iodine concentrate 8oz. dispensing bottle and diluent included to mix Iodine working solution.
A detailed product insert and convenient spout caps also included.