Gram color staining kit stabilised
Supplier: VWR International
89133-283
89133-283KT 113.75 USD
89133-283
Gram color staining kit stabilised
Gram color staining kit
Gram Stain Kit (stabilized Iodine) ready to use for the differentiation of bacteria
  • Set consists of "pre-mixed" iodine
  • For in vitro diagnostic use
  • Specifically formulatess for differential staining of bacteria from cultures or patient specimens
Kit includes one 8 oz. bottle of crystal violet, stabilized iodine, decolorizer, and safranin.
A detailed product insert and convenient spout caps also included.