Rapid Freeze Tissue Spray, Azer Scientific
Supplier: VWR International
89133-279
89133-279CS 1101.06 USD
89133-279
Rapid Freeze Tissue Spray, Azer Scientific
Freezing Sprays
This nonflammable, moisture-free spray is used to instantly cool or freeze samples for routine histology.
  • Nonflamable
  • Moisture-free
  • No CFCs or HFCs
  • 296 mL (10 oz.)
It can bring samples to –51°C (–60°F) in less than three seconds.
Rapid Freeze™ tissues spray contains no CFCs or HFCs and will not deplete stratospheric ozone.