Switchover System, Semi-Automatic, Diaphragm, Brass, Chrome Plated, Rotarex
Supplier: Ceodeux Inc.
TDL200125350
89133-268EA 4351.36 USD
89133-268
Switchover System, Semi-Automatic, Diaphragm, Brass, Chrome Plated, Rotarex
Gas Regulators
Ideally suited to insure continuous gas supply from many high pressure sources of high purity non-corrosive gases with low flow
Possible to manage 4 gas cylinders without any extension and a gas for purging operation (up to 6 cylinders without any extension - without using the purge line). Exists in a MANUAL, SEMI-AUTOMATIC and AUTOMATIC version.
When connected to a switch-over board or a supply board, optional alarms detect the moment when the cylinder is empty. They also indicate that the equipment works correctly. Visual and acoustic display. Repetition of the alarm by temporisation. Can be use with all kind of gauges. Connectable to remote alarms.
Dedicated to the supply of gas to analyzers and to the creation of controlled atmosphere in laboratories, control units, and for petrochemical applications.
Certifications: Alarms are CE marked (CEM directive).
Ordering information: System can be equipped with a collection tube on the safety relief valve and purge outlet, or with an outlet shut-off valve. Using contact gauges, the switch-over board can also be equipped with an alarm box to indicate the source status. Alarms do not include power supply cable.