Pig Blue® Barrel Top Absorbent Mat, New Pig
Supplier: New Pig
BLU255
89132-780CS 110.86 USD
89132-780
Pig Blue® Barrel Top Absorbent Mat, New Pig
Spill Control Materials
For your biggest pump and dispensing messes, put the power of highly-absorbent Pig Blue Mat to work
Contains 75% recycled fibers; Excellent in situations where green products are desired or required. Fire retardant for added safety; flame-tested in accordance with ASTM Standard E84-08.
Absorbs oils, coolants, solvents and water; not recommended for acids, bases or other corrosive liquids. Fits 55-gallon steel drum tops; pre-cut with holes for 3/4" and 2" bungs to provide a perfect fit. Dispenser box makes it easy to take, carry and store your mat while protecting it from dirt and moisture.
Soak up pump drips without cluttering up your work space with dirty rags. Consistent construction produces a mat that is free of light spots to absorb liquid evenly for better wicking and retention. FiberFusion Technology fuses superabsorbent natural fibers with strong synthetic fibers for one high-performance absorbent.