VWR® Premium Formalin Neutralizing Pads
Supplier: VWR International
89131-874
89131-874CS 2457.67 USD
89131-874
VWR® Premium Formalin Neutralizing Pads
Spill Control Materials
Durable, tear-resistant pads are manufactured with a bonded chemical that helps to minimize exposure to formaldehyde vapors.
  • Offensive Formalin Odors Eliminated
  • Disposable
  • Durable
  • Tear-Resistant
  • Blue Surface Provides Contrast
Pads immediately absorb and neutralize excess formalin run-off during dissection
They are also useful for lining shelves or transport containers where leakage may occur.
The distinctive blue surface of the pads offers an excellent contrast to even small-tissue specimens.