Heel Foot Grounder, Desco
Supplier: Desco
07590 07588 17202 17200
15551-110EA 16.46 USD
15551-110 89131-088 89129-466 89129-465
Heel Foot Grounder, Desco
Grounders Heel and Toe Grounders
Adjustable heel foot grounder with non-marring interior layer.
  • Non-marring interior layer
  • 1 meg and 2 meg resistors available
  • Made In The United States of America
Eight conductive strand nylon polyester grounding tab offers additional conductive material to provide a reliable connection with operator’s sweat-layer. Non-marring interior layer will not mar operator’s shoes. Resistor to protect operator from accidental contact with equipment line voltage.
Certifications: Suitable for ESD footwear component in flooring; Footwear System personnel grounding method (<3.5×107 ohms per ESD STM 97.1). Suitable for ESD footwear component in flooring; Footwear System personnel grounding method (<100 volts per ESD STM 97.2).