Eppendorf® PCR Cap Strips
Supplier: Eppendorf
0030124839
89130-978CS 123.83 USD
89130-978
Eppendorf® PCR Cap Strips
Tubes PCR Tubes
Cap strips provide easy and rapid sealing of Eppendorf® twin.tec PCR plates, PCR strips, and other standard multi-well plates.
  • Certified free of human DNA, DNase, RNase and PCR inhibitors
  • Autoclavable at 121 °C for 20 minutes
Strips feature eight microcaps designed for use with 0.2 ml wells. A tab at the end of each strip allows for easy removal.