Thermo Scientific™ MAS™ UA DipTube Controls, Thermo Scientific
Supplier: MICROGENICS CORPORATION (MAS)
Simplify the process of quality control during urinalysis testing with room-temperature, stable MAS™ UA DipTube Control
Key urinalysis test parameters provided in a single control. Tube capacity common within urinalysis test environment. Assayed for most major reagent strips and strip systems. Human urine matrix.
Analytes: Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells.
This liquid assayed urinalysis control for procedures used in routine urinalysis testing is conveniently-packed to afford immediate time and cost savings.
Ordering information: Not included, but requires: Slides, Pipetters, Centrifuge capable of generating an RCF of 400 or 450 for manual methods, Microscope with a 10x objective and a 40x objective lens, Chemical reagent strips.