Regulator, Line, Diaphragm, Stainless Steel, Rotarex
Supplier: Ceodeux Inc.
SI15116
89130-374EA 1670.18 USD
89130-374
Regulator, Line, Diaphragm, Stainless Steel, Rotarex
Gas Regulators
Used as a line regulator for high-flow industrial or lab applications.
As a second level of regulation the S 15 will supply a precise outlet pressure to the process. Can be used for wall or panel mounting with its compact design, the rear threads and fixing ring.
Pressure control with Balanced-Valve Technology: the effect of inlet pressure fluctuations on outlet pressure are minimized. BV-technology delivers an exceptionally stable outlet pressure and flow even with high flow line regulators. Longer useful regulator lifetime and lower total ownership cost.
Ordering information: Additional outlet pressures and flow rates available; contact your Avantor sales representative for more information.