Papanicolaou's solution EA 36
Supplier: VWR International
ES716
89130-180CS
308.6
USD
89130-180
Papanicolaou's solution EA 36
Papanicolaou's solution EA 36
These reagents work in conjunction to provide excellent cytoplasmic staining of gynecological samples.
- Excellent Cytoplasmic Staining of Gynecological and Non-Gynecological Samples
- Available in a Wide Range of Formulations
- Stains are Used in Conjunction with Hematoxylin Nuclear Stains in the Diagnosis of Malignant Cytological Disease
The range of formulations available allows the end user to select from various color intensities and hues.
Papanicolaou stains are used in conjunction with hematoxylin nuclear stains in the diagnosis of malignant cytological disease.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)