Papanicolaou's Orange G solution (OG 6) stain for cytology

Supplier: VWR International
Danger

ES713
89130-178CS 424.06 USD
89130-178
Papanicolaou's Orange G solution (OG 6) stain for cytology
Papanicolaou's Orange G solution (OG 6)

For routine cytology this counterstains will work with any hematoxylin for excellent cellular differentiation.


  • Pre-mixed and ready to use
  • Manufactured with certified dyes
  • Small batch production with long shelf life


OG-6 Pap Stain is also a cytoplasmic stain that stains target structures in cells different intensities of orange.

MDL Number: MFCD00147833
UN: 1170
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR