Transfection Reagent Diluent, 150 mM NaCl Solution, Polyplus-transfection
Supplier: Polyplus Transfection
201000002
89129-970EA 76.21 USD
89129-970
Transfection Reagent Diluent, 150 mM NaCl Solution, Polyplus-transfection
Transfection Reagents
This 150 mM NaCl solution is recommended as dilution reagent for DNA transfection with jetPEI®.