PULSin®, protein delivery reagent, Polyplus-transfection
Supplier: Polyplus Transfection
PULSin® protein delivery reagent is dedicated to the delivery of proteins, antibodies and peptides into cells and offers a powerful approach to functional studies
- Effective and robust delivery into the cytoplasm
- Delivery to a wide variety of cells including primary cells and Hard-to-transfect cells
- Highly efficient delivery
- Easy to use
PULSin® can be used to target intracellular proteins with antibodies in living cells without fixation.
Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Every batch of PULSin™ is tested by delivering R-phycoerythrin into HeLa cells.
Delivery information: Supplied with 20 µg R-Phycoerythrin (positive control) and Hepes dilution buffer. 0.4 ml of PULSin® protein delivery reagent is sufficient for 24 reactions in 6-well plates.