LockMailer™ Microscope Slide Jar, Simport Scientific
Supplier: Simport Scientific
M950-4MA
89129-506CS 324.25 USD
89129-506
LockMailer™ Microscope Slide Jar, Simport Scientific
Slide Mailers
Tamper-evident, multi-purpose container for mailing, staining, or storing microscope slides
  • Holds four standard microscope slides
  • Slotted container to keep slides separated
  • Optional unique tamper-evident cap
  • Different colors are available
Made of strong, clear polypropylene. It holds four standard slides vertically, and incorporates a unique tamper-evident, leakproof screw cap. Slots keep the slides from sticking together
Constructed of extra-strong and clear polypropylene. It will hold up to four standard 3×1" or 75×25 mm slides vertically. Inside channels are slotted to keep slides safely separated. Ideal for slide conveyors and specimen slide transport between a doctor's office and the lab.
The jar incorporates a unique tamper-evident leakproof screw cap, ensuring peace of mind during transport or storage situations where slides may have been manipulated without prior knowledge. Jar can also be used without the tamper-proof locking mechanism. For color coding purposes, use a Capinsert™ (see T345 series) that may be inserted on top of closure.
The container is designed for maximum stability on a benchtop while having an internal volume of only 12 ml.
Certifications: 95kPa tested.