CELLMASTER™ Roller Bottles, Polystyrene, Ribbed Surface, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
681075
89129-502CS 637.42 USD
89129-502 89501-646
CELLMASTER™ Roller Bottles, Polystyrene, Ribbed Surface, Sterile, Greiner Bio-One
Bottles Roller Bottles
Ribbed surface roller bottles come in long and short forms, and are made of high grade polystrene for stability and optical clarity, with physical surface treatment for improved cell adhesion.
  • Ribbed surface
  • High-grade polystyrene construction
  • HDPE screw cap construction with either standard seal or vented filter
  • Free of RNase, DNase, and human DNA
  • Non-cytotoxic (EN ISO 10993) and non-pyrogenic
Two-phase blow-molding procedure creates seamless construction so that liquids will not leak.
Screw caps are high-density polyethylene and come in standard seal (non-vented) or vented filter. Caps feature short threads and large knurls for improved grip and easy opening with a 2/3 turn. Standard seal cap sits flush with rim of bottle to prevent significant gas exchange. Vented filter cap guarantees tightly closed, contamination-free cultivation. Polyethylene terephthalate (PET) and polytetrafluorethane (PTFE) filter has hydrophobic capillary pore membrane, with defined pore size 0.2 µm. Filter provides optimal protection against contamination and a high gaseous exchange rate which enables mass cultivation of cells using biocarbonate buffer and CO₂.
Colors: clear roller bottle with black graduation print and white writing area, blue cap, white filter. Sterilized by irradiation (SAL 10⁻³) and tissue culture treated.
For single use only; lot number and best-before dates located on label for maximum traceability.
Certifications: USP Class VI certified.