HiPrep Phenyl FF (High Sub) Columns, Cytiva
Supplier: Cytiva
28-9365-45
89128-906EA 1102.08 USD
89128-906
HiPrep Phenyl FF (High Sub) Columns, Cytiva
Chromatography Columns
HiPrep Phenyl FF (High Sub) 16/10, are prepacked Phenyl Sepharose 6 Fast Flow columns ready to use for hydrophobic interaction chromatography (HIC).
  • 20 ml HIC columns provide fast, preparative separation of proteins and other biomolecules
  • Provide high capacity, excellent flow properties, and reliable, reproducible separations
  • Two levels of substitution help find optimal selectivity and binding capacity for a given application
  • Compatible with simple pump-based configurations or chromatography system such as ÄKTA design
The columns are designed for initial and intermediate purification steps, removing bulk impurities. Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.