HiPrep Butyl FF Columns, Cytiva
Supplier: Cytiva
28-9365-47
89128-902EA 1081.01 USD
89128-902
HiPrep Butyl FF Columns, Cytiva
Chromatography Columns
HiPrep Butyl FF (High Sub) 16/10, are prepacked Butyl Sepharose 4 Fast Flow columns ready to use for hydrophobic interaction chromatography (HIC).
  • 20 ml HIC columns provide fast, preparative separation of proteins and other biomolecules
  • Provide high capacity, excellent flow properties, and reliable, reproducible separations
  • Butyl Sepharose 4 Fast Flow is a well-established, standard aliphatic HIC medium for capture and intermediate purification
  • Compatible with simple pump-based configurations or chromatography system such as ÄKTA design
The columns are designed for initial and intermediate purification, removing bulk impurities.