HiPrep Q FF Columns, Cytiva
Supplier: Cytiva
28-9365-43
89128-890EA 1023.16 USD
89128-890
HiPrep Q FF Columns, Cytiva
Chromatography Columns
HiPrep Q FF is a strong anion exchanger prepacked with Q Sepharose Fast Flow, ready-to-use for fast, preparative separations of proteins and other biomolecules using ion exchange (IEX) chromatography.
  • HiPrep columns provide fast, reliable, and reproducible preparative ion exchange separations
  • Good flow rates and high capacity
  • The industry standard for ion exchange chromatography
Compatible with single-pump configurations and chromatography systems such as ÄKTA design. Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient.