HiPrep SP FF Columns, Cytiva
Supplier: Cytiva
28-9365-44
89128-886EA 1002.97 USD
89128-886
HiPrep SP FF Columns, Cytiva
Chromatography Columns
HiPrep SP FF is a strong cation exchanger prepacked with SP Sepharose fast flow, ready-to-use for fast, preparative separations of proteins and other biomolecules using ion exchange (IEX) chromatography.
  • HiPrep columns provide fast, reliable, and reproducible preparative ion exchange separations
  • Good flow rates and high capacity
  • The industry standard for ion exchange chromatography
  • Compatible with single-pump configurations and chromatography systems such as ÄKTA design
  • Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient