HiPrep DEAE FF Columns, Cytiva
Supplier: Cytiva
28-9365-41
89128-880EA 1047.65 USD
89128-880
HiPrep DEAE FF Columns, Cytiva
Chromatography Columns
HiPrep DEAE FF 16/10 is a weak anion exchanger prepacked with DEAE Sepharose Fast Flow, ready-to-use for fast, preparative separations of proteins and other biomolecules using ion exchange (IEX) chromatography.
  • 20 ml HiPrep columns provide fast, reliable, and reproducible preparative ion exchange separations
  • Good flow rates and high capacity
  • The industry standard for ion exchange chromatography
  • Compatible with single-pump configurations and chromatography systems such as ÄKTA design
  • Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient
It provides fast, preparative separations of proteins and other biomolecules.