Basic fuchsin in alcohol reagent A for PREVI® COLOR GRAM instrument

Supplier: bioMerieux
Danger

Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline

29585
89127-612EA 98.94 USD
89127-612
Basic fuchsin in alcohol reagent A for PREVI® COLOR GRAM instrument
Basic Fuchsin

RA4 FUCHSIN solution


  • Easy to use
  • Premade reagents
  • No mess in the lab


The PREVI® Color Gram dyes are used to stain micro-organisms according to Gram’s method. They are intended for use exclusively with the PREVI® Color Gram instrument.

Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
MDL Number: MFCD00012569
CAS Number: 632-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR