Transfer Assemblies, Sterile, Advanced Scientifics
Supplier: Advanced Scientific, Inc.
SS00042-I SS00043-I SS00044-I SS00045-I SS00062-I SS00063-I SS00064-I SS00065-I
89127-084CS 7287.3 USD
89127-084 89127-086 89127-088 89127-090 89127-092 89127-094 89127-096 89127-098
Transfer Assemblies, Sterile, Advanced Scientifics
Sterile silicone tubing sets to ease the transfer of bulk fluids from one container to another.
Ideal for biopharmaceutical, laboratory, and diagnostic applications.
Delivery information: Sets include 91.4cm (36") tubing and pinch clamp.