Pillow-Style Tank Liners, Advanced Scientifics
Supplier: Advanced Scientific, Inc.
B100038-I B100037-I B100049-I B100050-I B100053-I B100054-I B100048-I
89127-002CS 270.65 USD
89127-002 89127-004 89127-006 89127-008 89127-014 89127-016 89127-001
Pillow-Style Tank Liners, Advanced Scientifics
Linear low-density polyethylene tank liners are manufactured in an ISO Class 7 (FED-STD-209E Class 10,000/M5.5) cleanroom environment, packaged clean, irradiated, and delivered sterile.
Economical solution for cleaning and turnover concerns associated with reusable stainless tanks.