Lindstrom® Diagonal Flush Cutter, Oval Head, JH Williams Tool Group
Supplier: J.H. Williams Tool Group (Lindstrom)
8141
89125-250EA 528.25 USD
89125-250
Lindstrom® Diagonal Flush Cutter, Oval Head, JH Williams Tool Group
Cutters
Lindstrom 80-Series has precision-forged and crafted oval head cutters.
  • Precision induction hardened edges 63 - 65 HR(c)
  • Carbon/chrome ball bearing steel 1%
  • Replaceable springs and precision screw joint
  • CNC machining
  • ESD safe handles
No ergonomic handle.