CiDehol® Filtered 60% IPA Solution, Decon Labs
Supplier: Decon Labs
8800
89125-216CS 339.93 USD
89125-216
CiDehol® Filtered 60% IPA Solution, Decon Labs
Cleaning Agents Disinfectants
This 60% isopropyl alcohol (IPA) solution is filtered to 0.2µm to remove particulate contamination
Ideal for filter integrity and testing and for cleaning labs, production areas, and equipment.