Media Holder for Printers - External Stand, Brady
Supplier: Brady Worldwide
114540
89125-024EA 107.92 USD
89125-024
Media Holder for Printers - External Stand, Brady
Label Holders
This External Media Holder fits 3" core label rolls for high-volume printing.
  • Package includes media holder only (printer and label rolls sold separately)
  • Saves time and increases efficiency for projects requiring high-volume printing
  • Increases the range of materials that are compatible with the printer
The External Media Holder attaches to the back of the printer, enabling users to run 3" core rolls with more labels per roll for high volume printing.