Axygen® Robotic Tips for Corbett X-tractor, Corning
Supplier: Corning
CTF-199-R-S
89125-000CS 514.39 USD
89125-000
Axygen® Robotic Tips for Corbett X-tractor, Corning
Pipette Tips
Tips in 96-well format designed for Corbett X-tractor Gene 8 channel head.
  • Clear polypropylene
  • With aerosol-resistant filters
  • Optimised fine-point for accurate dispensing
  • Certified RNase, DNase and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.