Axygen® Robotic Pipette Tips, ZT Series, Corning
Supplier: Corning
ZT-384-C-R
89124-996CS 5619.88 USD
89124-996
Axygen® Robotic Pipette Tips, ZT Series, Corning
Pipette Tips
The ZT Series Robotic tips for the Zymark® RapidPlate/Cyclone automated systems are certified RNase-, DNase-, and pyrogen-free.
The ZT Series Robotic tips are engineered to exact dimensions of OEM tips to ensure automation compatibility.