MBP Molecular Devices® -Style 384-Well Tips, Thermo Scientific
Supplier: Thermo Scientific
931-261
89108-097CS 5362.33 USD
89108-097
MBP Molecular Devices® -Style 384-Well Tips, Thermo Scientific
Pipette Tips
MBP leads the way in FLIPR style tips with our SoftFit technology.
  • Thin wall design that creates a better seal, reduces the loading force required
  • Improves %CVs, and reduces wear and tear on the machine
ART is a registered trademark of MBP and is protected by U.S. Patent # 5,156,811. SoftFit is a trademark of Molecular Bio-Product. Molecular Devices and FLIPR are registered trademarks of Molecular Devices Corp.