Virtua™ Protective Eyewear, 3M™
Supplier: 3M
7100112434 7100109223 7100123356 7010386340 7100114652
47746-237CS 84.46 USD
47746-237 89006-547 47746-233 47746-241 89107-432 89217-718
Virtua™ Protective Eyewear, 3M™
Safety Glasses
Virtua™ Protective Eyewear provides unisex styling, protection, and affordability for site visitors or single-use protection.
  • Designed for indoor or outdoor use
  • Wide profile provides necessary impact protection and an unobstructed view
Lightweight, unisex Virtua™ Protective Eyewear combines versatility and value with high wraparound coverage, polycarbonate construction, and a hard coat, scratch-resistant lens for a comfortable, contoured fit.
Certifications: ANSI Z87.1.