Nunc® IVF Centerwell Dish, Thermo Scientific
Supplier: Thermo Fisher Scientific
150260
89106-784CS 420.7 USD
89106-784
Nunc® IVF Centerwell Dish, Thermo Scientific
Culture Dishes Petri Dishes
Thermo Scientific Nunc® provide an extensive portfolio of products intended for use in human In Vitro Fertilization (IVF)
The Thermo Scientific Nunc® IVF Center Well Dish features a perfect angle of reservoir well walls for locating and easy washing of samples. It maintains optimal temperature with a true flat bottom, while providing increased safety with the easy grip design.
Thermo Scientific Nunc® InVitro Fertilization Products provide security, safety and reproducibility for your valuable samples, and are uniquely certified based on a 1-cell stage mouse embryo toxicity test. Stringent lot-level manufacturing controls ensure viability, reliability and reproducibility. Lid is Present.
Products are produced under ISO 13485, MEA tested, CE marked class II medical device and FDA cleared for use in IVF.
Certifications: CE and FDA