VWR® Tape and Label Dispenser
Supplier: VWR International
89098-146
89098-146CS 22.48 USD
89098-146
VWR® Tape and Label Dispenser
Label and Tape Dispensers
This durable plastic handheld dispenser for tapes and labels accommodates a maximum width of 2.9 cm (11/8").
  • Durable plastic construction
  • Locking clip
  • Dispenses a range of label sizes
Easily dispenses 12.7m (41.7') long rolls of tape in widths of 12.7 mm (1/2"), 19 mm (3/4"), or 25.4 mm (1"), on a 25.4 mm (1") core. Includes two spacers and a plastic locking clip.
Ordering information: Tape and labels sold separately.