VWR® Slide/Test Tube Labels
Supplier: VWR International
89097-863
89097-863CS 37.82 USD
89097-863
VWR® Slide/Test Tube Labels
Labels
Clear, self-laminating slide/test tube labels feature a 2.4×2.4 cm (¹⁵/₁₆ ×¹⁵/₁₆") white UV square for identification purposes.
  • Clear design
  • Self-laminating labels
  • Suitable for thermal transfer printers
  • May be written by hand
  • White UV identification square
The wrap around design protects data from stains, reagents, and xylene.
Ordering information: Labels are supplied 1400 per roll (1 roll per case).