VWR® Patient ID Labels
Supplier: VWR International
89097-834
89097-834CS 45.26 USD
89097-834
VWR® Patient ID Labels
Labels
Smudge-proof flexo labels feature a removeable adhesive for patient ID labeling.
  • Multiple lines for identification
  • Smudge-proof
  • Carrier width is 82.5 mm
All labels allow the user to dial in two lines of information with up to eight characters per line to be printed. Available characters include two Gregorian dates for printing expiration and autoclave dating with an additional band for load and sterilizer numbers.