Jacketed Pilot Plant Reactor Pressure Relief Manifold, Ace Glass Incorporated
Supplier: Ace Glass
10015-10
89097-262EA 4437.56 USD
89097-262
Jacketed Pilot Plant Reactor Pressure Relief Manifold, Ace Glass Incorporated
Manifolds
A pressure relief manifold for a jacketed, reaction vessel is designed to protect the vessel from breakage while isolated or disconnected from the circulator
Installed on the outlet side of the vessel,the manifold will reduce the liklihood of breakage caused by an excessive pressure differential exerted by the thermal transfer fluid. Complete system includes aluminum body, outlet fittings, hose, relief valve set at 10 psi, and pressure gauge. Fits 10 l and above.