VWR® TraceClean® 20 ml Vials
Supplier: VWR International
QLFV15900002 QLFV15900026 QLFV15900008 QLFV15900020 QLFV15900032 QLFV15900014
89093-830CS 245.91 USD
89093-830 89093-838 89093-848 89093-854 89093-860 89096-774
VWR® TraceClean® 20 ml Vials
Vials
Type I clear borosilicate glass vials are for use in various analyses and in specimen, extract, and standards storage.
  • Borosilicate glass fabrication
  • Available with polypropylene solid-top or open-top caps
  • Prepared to avoid volatile contamination
They are prepared in a solvent-free facility to prevent possible volatile contamination
Processed (PC) and Quality-Assured® (QA) vials follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA vials include a Certificate of Analysis in every case. QA vials also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) vials do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Vials are available with polypropylene solid-top or open-top caps. Solid-top caps feature fluoropolymer resin liners. Open-top caps feature ultrasonically bonded 3.1 mm (¹/₈") fluoropolymer resin/silicone septa and are for volatile organic analyses.