Acetonitrile ≥99.5% (by GC-FID) ACS

Supplier: Tedia
Danger

AR0116-008
89096-138EA 3101.39 USD
89096-138
Acetonitrile ≥99.5% (by GC-FID) ACS
Acetonitrile

  • Meets reagent specifications for testing USP/NF monographs.
  • Packaged under Nitrogen
  • Submicron filtered

Formula: H₃CCN
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% min.
Color (APHA) ≤10
Residue after evaporation ≤0.005%
Titrable acid ≤0.008meq/g
Titrable base ≤0.0006meq/g
Water ≤0.3%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR