VWR® 10/30 Ground Joint Thermometers
Supplier: VWR International
89095-788 89095-790 89095-794 89095-796 89095-798
89095-788CS 1488.92 USD
89095-788 89095-790 89095-794 89095-796 89095-798
VWR® 10/30 Ground Joint Thermometers
Thermometers Analog Thermometers
Thermometers feature a 10/30 ground joint and Celsius scale.
  • Individually serialized
  • Contain red organic liquid against white back glass
Each thermometer is calibrated for specific immersion depths.