Translucent UN Rated Poly Pail, New Pig
Supplier: New Pig
DRM282
89095-316EA 66.72 USD
89095-316
Translucent UN Rated Poly Pail, New Pig
Buckets
Translucent walls eliminate guesswork to help prevent overfilling or running out of materials
  • Walls eliminate guesswork to help prevent overfilling or running out of materials
  • Made from FDA-approved resins for containment of food products
  • High-density polyethylene construction is strong and lightweight
  • 5-Gallon pail provides efficient storage and transfer of smaller quantities
  • Sturdy fold-down handle makes pail easy to carry
  • UN1H1/Y1.8/100
Made from FDA Approved resins for containment of food products. High-density polyethylene construction is strong and light-weight. 5-gallon pail provides efficient storage and transfer of smaller quantities. Sturdy fold-down handle makes pail easy to carry. UN1H1/Y1.8/100.