PIG® Green Ham-O® Absorbent Mat Pad in Dispenser Box, New Pig
Supplier: New Pig
MAT268
89095-166EA 293.9 USD
89095-166
PIG® Green Ham-O® Absorbent Mat Pad in Dispenser Box, New Pig
Spill Control Materials
Exclusive "hamouflage" top layer saves on product and disposal costs; absorbed grime is invisible, so you'll leave mat in place longer and save money
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups.
Fast-wicking "diaper action" transfers liquids from the top layer into the absorbent inner layers; spills disappear in seconds. Tough, spunbond top layer stands up to heavy foot traffic; won't rip, tear, fray or create a trip hazard, even when saturated. Under the Ham-O top layer is the proven eight-layer, 100% polypropyleneMore Product Information